4,4'-Biphenol 97%

Supplier: Ambeed
Danger

Synonyms: [1,1'-Biphenyl]-4,4'-diol, 4,4'-Dihydroxybiphenyl, Biphenyl-4,4'-diol

A337203-100G A337203-10G A337203-500G A337203-1000G A337203-25G
103810-164EA 74 USD
103810-164 76803-090 76803-092 76803-094 76839-716 76839-718
4,4'-Biphenol 97%
4,4'-Biphenol

Precaución: for Research Use Only

Formula: C₁₂H₁₀O₂
MW: 186.21 g/mol
Melting Pt: 280…282 °C
Flash Pt: 176.1 °C
Storage Temperature: Ambient
MDL Number: MFCD00002348
CAS Number: 92-88-6
UN: 3077
ADR: 9,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR