4,4'-Dihydroxybenzophenone 97%

Supplier: Ambeed
Warning

A315950-1000G A315950-25G A315950-5G A315950-100G A315950-500G
103810-090EA 135 USD
103810-090 103836-012 76838-846 76838-848 76838-850 77114-662 77114-664
4,4'-Dihydroxybenzophenone 97%
4,4'-Dihydroxybenzophenone

Precaución: for Research Use Only

Formula: C₁₃H₁₀O₃
Boiling Pt: 444.8 °C (760 mm Hg)
Melting Pt: 213…215 °C
Flash Pt: 237 °C
Storage Temperature: Ambient
MDL Number: MFCD00002358
CAS Number: 611-99-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR