4,4-Difluoropiperidine hydrochloride 97%

Supplier: Ambeed
Warning

Synonyms: 4,4-Difluoropiperidine HCl

A331925-1G A331925-5G A331925-100G A331925-10G A331925-250MG A331925-25G
103825-592EA 6 USD
103825-592 103836-560 76839-552 76839-554 77167-442 77167-444 77167-446 77167-448
4,4-Difluoropiperidine hydrochloride 97%
4,4-Difluoropiperidine hydrochloride

Precaución: for Research Use Only

Formula: C₅H₁₀ClF₂N
MW: 157.59 g/mol
Melting Pt: 173…177 °C
Storage Temperature: Refrigerator
MDL Number: MFCD03095381
CAS Number: 144230-52-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR