4,4-Difluoropiperidine 97%

Supplier: Ambeed
Warning

A410500-1G A410500-5G A410500-100MG A410500-10G A410500-250MG A410500-25G
103827-214EA 8 USD
103827-214 103838-262 76841-782 76841-784 77291-286 77291-288 77291-290 77291-292
4,4-Difluoropiperidine 97%
4,4-Difluoropiperidine

Precaución: for Research Use Only

Formula: C₅H₉F₂N
Boiling Pt: 114.3 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD03094281
CAS Number: 21987-29-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR