4,4-Difluorocyclohexanone 98%

Supplier: Ambeed
Warning

A212240-1G A212240-25G A212240-5G A212240-100MG A212240-10G A212240-100G A212240-250MG
103818-176EA 16 USD
103818-176 103848-876 77087-328 77087-330 77087-332 77287-990 77287-992 77748-321 77748-322
4,4-Difluorocyclohexanone 98%
4,4-Difluorocyclohexanone

Precaución: for Research Use Only

Formula: C₆H₈F₂O
Boiling Pt: 157.4 °C (760 mm Hg)
Melting Pt: 35…36 °C
Storage Temperature: Freezer
MDL Number: MFCD08146635
CAS Number: 22515-18-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR