4,4-Difluorocyclohexanol 95%

Supplier: Ambeed
Warning

A279076-100MG A279076-10G A279076-250MG A279076-1G
103813-450EA 16 USD
103813-450 103853-394 76787-364 76787-366 76787-368 77093-320
4,4-Difluorocyclohexanol 95%
4,4-Difluorocyclohexanol

Precaución: for Research Use Only

Formula: C₆H₁₀F₂O
MW: 136.1398 g/mol
Melting Pt: 61…63 °C
Storage Temperature: Refrigerator
MDL Number: MFCD10000566
CAS Number: 22419-35-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR