4,4-Difluorocyclohexanecarboxylic acid 97%

Supplier: Ambeed
Warning

Synonyms: 4,4-difluorocyclohexane-1-carboxylic acid

A394947-25G A394947-100G
103826-952EA 9 USD
103826-952 103837-990 76841-412 77003-796
4,4-Difluorocyclohexanecarboxylic acid 97%
4,4-Difluorocyclohexanecarboxylic acid

Precaución: for Research Use Only

Formula: C₇H₁₀F₂O₂
MW: 164.15 g/mol
Melting Pt: 103…107 °C
Flash Pt: 99.6 °C
Storage Temperature: Ambient
MDL Number: MFCD03788493
CAS Number: 122665-97-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR