4,4-Difluorocyclohexanecarboxaldehyde 95%

Supplier: Ambeed
Warning

Synonyms: 4,4-Difluorocyclohexanecarbaldehyde

A131297-250MG A131297-1G A131297-25G
103801-560EA 53 USD
103801-560 76688-634 77314-054 77624-112
4,4-Difluorocyclohexanecarboxaldehyde 95%
4,4-Difluorocyclohexanecarboxaldehyde

Precaución: for Research Use Only

Formula: C₇H₁₀F₂O
MW: 148.15 g/mol
Boiling Pt: 165 °C (760 mm Hg)
Storage Temperature: Freezer
MDL Number: MFCD09263468
CAS Number: 265108-36-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR