4,4'-Dichlorobenzophenone 98%

Supplier: Ambeed
Warning

Synonyms: 4,4-Dichlorobenzophenone, 4,4-DBP, Bis(4-chlorophenyl) ketone

A121760-100G A121760-10G A121760-25G A121760-500G A121760-5G A121760-1000G
76870-248EA 20.16 USD
76870-248 76870-250 76870-252 76870-254 77297-846 77566-264
4,4'-Dichlorobenzophenone 98%
4,4'-Dichlorobenzophenone

Precaución: For research use only.

Formula: C₁₃H₈Cl₂O
MW: 251.11 g/mol
Boiling Pt: 353 °C (760 mm Hg)
Melting Pt: 144…147 °C at N/A mmHg
Flash Pt: 352 °C
Storage Temperature: Ambient
MDL Number: MFCD00000623
CAS Number: 90-98-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR