4,4'-Dibromobenzophenone 98%
Supplier: Ambeed
|
Warning
|
A156486-100G
A156486-10G
A156486-500G
A156486-1G
A156486-25G
A156486-5G
103804-894EA
7
USD
103804-894
103843-094
77286-286
77286-288
77286-290
77300-192
77318-876
77318-878
4,4'-Dibromobenzophenone 98%
4,4'-Dibromobenzophenone
Precaución: for Research Use Only
Formula:
C₁₃H₈Br₂O Boiling Pt: 395.6 °C (760 mm Hg) Melting Pt: 171…174 °C Flash Pt: 121 °C Storage Temperature: Ambient |
MDL Number:
MFCD00016330 CAS Number: 3988-03-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...