4,4'-Dibromobenzil 97%

Supplier: Ambeed
Warning

A212124-100G A212124-10G A212124-1G A212124-25G A212124-5G
103818-168EA 11 USD
103818-168 103848-868 76785-714 76785-716 77303-224 77303-226 77303-228
4,4'-Dibromobenzil 97%
4,4'-Dibromobenzil

Precaución: for Research Use Only

Formula: C₁₄H₈Br₂O₂
Boiling Pt: 457.2 °C (760 mm Hg)
Melting Pt: 224…226 °C
Flash Pt: 142.4 °C
Storage Temperature: Ambient
MDL Number: MFCD00000104
CAS Number: 35578-47-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR