4,4''-Dibromo-p-terphenyl 97%

Supplier: Ambeed
Warning

Synonyms: 4,4''-Dibromo-1,1':4',1''-terphenyl

A205295-1G A205295-5G A205295-10G A205295-250MG A205295-25G
103817-622EA 8 USD
103817-622 103848-290 77086-498 77086-500 77112-302 77112-304 77112-306
4,4''-Dibromo-p-terphenyl 97%
4,4''-Dibromo-p-terphenyl

Precaución: for Research Use Only

Formula: C₁₈H₁₂Br₂
Boiling Pt: 481.7 °C (760 mm Hg)
Melting Pt: 315 °C
Storage Temperature: Ambient
MDL Number: MFCD00272889
CAS Number: 17788-94-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR