4,4'-Dibromo-[1,1'-biphenyl]-2,2'-diamine 97%

Supplier: Ambeed
Danger

A642349-10G A642349-250MG A642349-1G A642349-25G
103840-348EA 76 USD
103840-348 77018-294 77018-296 77308-046 77308-048
4,4'-Dibromo-[1,1'-biphenyl]-2,2'-diamine 97%
4,4'-Dibromo-[1,1'-biphenyl]-2,2'-diamine

Precaución: for Research Use Only

Formula: C₁₂H₁₀Br₂N₂
Melting Pt: 113…118 °C
Storage Temperature: Ambient
MDL Number: MFCD14582981
CAS Number: 136630-36-9
UN: 3077
ADR: 9,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR