4,4''-Diamino-p-terphenyl 95%

Supplier: Ambeed
Warning

A160215-10G A160215-250MG A160215-25G A160215-1G A160215-5G
103805-348EA 14 USD
103805-348 103843-578 77150-834 77150-836 77150-838 77300-448 77300-450
4,4''-Diamino-p-terphenyl 95%
4,4''-Diamino-p-terphenyl

Precaución: for Research Use Only

Formula: C₁₈H₁₆N₂
MW: 260.34 g/mol
Boiling Pt: 484.2 °C (760 mm Hg)
Melting Pt: 242…244 °C
Flash Pt: 296.3 °C
Storage Temperature: Ambient
MDL Number: MFCD00051532
CAS Number: 3365-85-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR