4,4'-Diamino-[1,1'-biphenyl]-2,2'-dicarboxylic acid 95%

Supplier: Ambeed
Warning

A361510-5G A361510-10G A361510-250MG
103826-286EA 49 USD
103826-286 103837-300 76840-514 77290-774 77290-776
4,4'-Diamino-[1,1'-biphenyl]-2,2'-dicarboxylic acid 95%
4,4'-Diamino-[1,1'-biphenyl]-2,2'-dicarboxylic acid

Precaución: for Research Use Only

Formula: C₁₄H₁₂N₂O₄
Boiling Pt: 515.1 °C (760 mm Hg)
Flash Pt: 265.3 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00156997
CAS Number: 17557-76-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR