4,4'-Biphenyldimethanol 98%

Supplier: Ambeed
Warning

A389988-1G A389988-25G A389988-5G A389988-250MG A389988-100G A389988-10G
103826-858EA 23 USD
103826-858 103837-900 76841-278 76841-280 76841-282 77003-742 77601-714 77672-938
4,4'-Biphenyldimethanol 98%
4,4'-Biphenyldimethanol

Precaución: for Research Use Only

Formula: C₁₄H₁₄O₂
Boiling Pt: 416.3 °C (760 mm Hg)
Melting Pt: 191…192 °C
Flash Pt: 204.6 °C
Storage Temperature: Ambient
MDL Number: MFCD00016713
CAS Number: 1667-12-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR