4,4'-Bipyridyl 98%

Supplier: Ambeed
Danger

Synonyms: 4,4'-Bipyridine, 4,4'-Dipyridyl

A157160-10G A157160-25G A157160-5G
103808-848EA 248 USD
103808-848 103843-176 76973-362 77300-238 77300-240
4,4'-Bipyridyl 98%
4,4'-Bipyridyl

Precaución: for Research Use Only

Formula: C₁₀H₈N₂
MW: 156.19 g/mol
Boiling Pt: 304.8 °C (760 mm Hg)
Melting Pt: 109…112 °C
Flash Pt: 304 °C
Storage Temperature: Ambient
MDL Number: MFCD00006416
CAS Number: 553-26-4
UN: 2811
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR