Phenylboronic acid pinacol ester 95%
Supplier: Ambeed
|
Warning
|
Synonyms:
(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzene, Benzeneboronic acid, pinacol ester, 4,4,5,5-Tetramethyl-2-phenyl-1,3,2-dioxaborolane, 2-Phenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
A105502-100G
A105502-10G
A105502-25G
A105502-500G
A105502-5G
103822-626EA
5
USD
103822-626
76717-780
76717-782
76717-784
76717-786
77309-494
Phenylboronic acid pinacol ester 95%
Phenylboronic acid pinacol ester
Precaución: for Research Use Only
Formula:
C₁₂H₁₇BO₂ Boiling Pt: 282.1 °C (760 mm Hg) Melting Pt: 27 °C Flash Pt: 124.4 °C Storage Temperature: Ambient |
MDL Number:
MFCD00966985 CAS Number: 24388-23-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...