3-Methylphenylboronic acid pinacol ester 98%

Supplier: Ambeed
Warning

Synonyms: 3-Methylbenzeneboronic acid, pinacol ester, 4,4,5,5-Tetramethyl-2-(m-tolyl)-1,3,2-dioxaborolane, 3-Tolylboronic acid pinacol ester, m-Tolylboronic acid Pinacol ester, 3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)toluene

A145191-10G A145191-250MG A145191-1G A145191-25G A145191-5G
103803-502EA 6 USD
103803-502 103841-584 76719-480 76719-482 77316-862 77316-864 77316-866
3-Methylphenylboronic acid pinacol ester 98%
3-Methylphenylboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₃H₁₉BO₂
Boiling Pt: 307.2 °C (760 mm Hg)
Storage Temperature: Ambient
MDL Number: MFCD05864303
CAS Number: 253342-48-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR