4,4,5,5-Tetramethyl-2-(5-methylthiophen-2-yl)-1,3,2-dioxaborolane 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
5-Methylthiophen-2-boronic acid pinacol ester
A164313-10G
A164313-25G
A164313-1G
A164313-5G
103805-842EA
9
USD
103805-842
103844-102
76768-754
77300-706
77320-260
77320-262
4,4,5,5-Tetramethyl-2-(5-methylthiophen-2-yl)-1,3,2-dioxaborolane 97%
4,4,5,5-Tetramethyl-2-(5-methylthiophen-2-yl)-1,3,2-dioxaborolane
Precaución: for Research Use Only
Formula:
C₁₁H₁₇BO₂S MW: 224.12 g/mol Boiling Pt: 308.2 °C (760 mm Hg) Flash Pt: 105 °C Storage Temperature: Ambient |
MDL Number:
MFCD05664108 CAS Number: 476004-80-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...