2,2,6,6-Tetramethyl-3,6-dihydro-2H-pyran-4-boronic acid pinacol ester 95%

Supplier: Ambeed
Warning

Synonyms: 2,2,6,6-Tetramethyl-4-(4,4,5,5-tetramethyl-[1,3,2]dioxaborolan-2-yl)-3,6-dihydro-2H-pyran, (2,2,6,6-Tetramethyl-3,6-dihydro-2H-pyran-4-yl)boronic acid pinacol ester, 4,4,5,5-tetramethyl-2-(2,2,6,6-tetramethyl-3,6-dihydro-2H-pyran-4-yl)-1,3,2-dioxaborolane

A316454-100MG A316454-250MG
103825-106EA 84 USD
103825-106 103836-032 76816-976 76816-978
2,2,6,6-Tetramethyl-3,6-dihydro-2H-pyran-4-boronic acid pinacol ester 95%
2,2,6,6-Tetramethyl-3,6-dihydro-2H-pyran-4-boronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₅H₂₇BO₃
MW: 266.19 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD11044439
CAS Number: 1142363-56-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR