4,4,5,5-Tetramethyl-2-(1,4-dioxaspiro[4.5]dec-7-en-8-yl)-1,3,2-dioxaborolane 97%

Supplier: Ambeed
Warning

Synonyms: 8-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-1,4-dioxaspiro[4.5]dec-7-ene, 4,4,5,5-Tetramethyl-2-(1,4-dioxaspiro[4.5]dec-7-en-8-yl)-1,3,2-dioxaborolane, 1,4-Dioxaspiro[4,5]dec-7-en-8-boronic acid Pinacol ester

A215680-250MG A215680-1G A215680-25G A215680-5G A215680-10G A215680-100G
103818-508EA 8 USD
103818-508 103849-216 76785-802 77087-756 77087-758 77087-760 77568-084 77593-406
4,4,5,5-Tetramethyl-2-(1,4-dioxaspiro[4.5]dec-7-en-8-yl)-1,3,2-dioxaborolane 97%
4,4,5,5-Tetramethyl-2-(1,4-dioxaspiro[4.5]dec-7-en-8-yl)-1,3,2-dioxaborolane

Precaución: For research use only.

Formula: C₁₄H₂₃BO₄
MW: 266.1410 g/mol
Melting Pt: 60…62 °C
Storage Temperature: Freezer
MDL Number: MFCD04115656
CAS Number: 680596-79-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR