4,4,5,5-Tetramethyl-2-(1,2,3,6-tetrahydro-[1,1'-biphenyl]-4-yl)-1,3,2-dioxaborolane 95%
Supplier: Ambeed
|
Warning
|
A516688-1G
A516688-5G
A516688-100MG
A516688-250MG
103828-458EA
158
USD
103828-458
103839-564
76843-552
76843-554
76979-978
76979-980
4,4,5,5-Tetramethyl-2-(1,2,3,6-tetrahydro-[1,1'-biphenyl]-4-yl)-1,3,2-dioxaborolane 95%
4,4,5,5-Tetramethyl-2-(1,2,3,6-tetrahydro-[1,1'-biphenyl]-4-yl)-1,3,2-dioxaborolane
Precaución: For research use only
Formula:
C₁₈H₂₅BO₂ MW: 284.2 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD11520534 CAS Number: 287944-05-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...