4,4,4-Trifluorobutyric acid 98%

Supplier: Ambeed
Danger

A799080-25G A799080-250MG
103841-056EA 14 USD
103841-056 76845-300 77293-784
4,4,4-Trifluorobutyric acid 98%
4,4,4-Trifluorobutyric acid

Precaución: for Research Use Only

Formula: C₄H₅F₃O₂
MW: 142.08 g/mol
Boiling Pt: 149.1 °C (760 mm Hg)
Melting Pt: 30 °C
Flash Pt: 44 °C
Storage Temperature: Ambient
MDL Number: MFCD00077604
CAS Number: 406-93-9
UN: 3261
ADR: 8,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR