4,4',4''-Triaminotriphenylamine 97%

Supplier: Ambeed
Warning

Synonyms: Tris(4-aminophenyl)amine

A208803-100G A208803-10G A208803-250MG A208803-1G A208803-25G A208803-5G
103817-886EA 15 USD
103817-886 103848-574 76975-440 76975-442 76975-444 77086-902 77086-904 77086-906
4,4',4''-Triaminotriphenylamine 97%
4,4',4''-Triaminotriphenylamine

Precaución: for Research Use Only

Formula: C₁₈H₁₈N₄
MW: 290.38 g/mol
Melting Pt: 230 °C
Flash Pt: 294.1 °C
Storage Temperature: Ambient
MDL Number: MFCD00778301
CAS Number: 5981-09-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR