4,4',4''-Nitrilotribenzoic acid 95%

Supplier: Ambeed
Warning

A166116-250MG A166116-1G A166116-25G A166116-5G
103806-016EA 27 USD
103806-016 103844-272 77236-108 77320-502 77320-504 77320-506
4,4',4''-Nitrilotribenzoic acid 95%
4,4',4''-Nitrilotribenzoic acid

Precaución: for Research Use Only

Formula: C₂₁H₁₅NO₆
MW: 377.35 g/mol
Boiling Pt: 690.0±50.0 °C (760 mm Hg)
Melting Pt: >320 °C
Flash Pt: 371.1 °C
Storage Temperature: Ambient
MDL Number: MFCD22123674
CAS Number: 118996-38-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR