Tetrakis(4-aminophenyl)methane 95%

Supplier: Ambeed
Warning

Synonyms: 4,4',4'',4'''-Methanetetrayltetraaniline

A101665-250MG A101665-25G A101665-1G A101665-5G A101665-10G
103796-078EA 24 USD
103796-078 103821-774 76982-186 76982-188 77282-790 77282-792 77610-748
Tetrakis(4-aminophenyl)methane 95%
Tetrakis(4-aminophenyl)methane

Precaución: for Research Use Only

Formula: C₂₅H₂₄N₄
MW: 380.5 g/mol
Melting Pt: 334…339 °C
Storage Temperature: Ambient
MDL Number: MFCD12546930
CAS Number: 60532-63-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR