Bis(hexylene glycolato)diboron 98%

Supplier: Ambeed
Warning

Synonyms: 4,4,4',4',6,6'-Hexamethyl-2,2'-bi-1,3,2-dioxaborinane, 4,4,4',4',6,6'-Hexamethyl-2,2'-bi(1,3,2-dioxaborinane)

A223271-100G A223271-10G A223271-1G A223271-25G A223271-5G
103819-026EA 5 USD
103819-026 103849-908 76722-022 76722-024 77088-522 77088-524 77088-526
Bis(hexylene glycolato)diboron 98%
Bis(hexylene glycolato)diboron

Precaución: for Research Use Only

Formula: C₁₂H₂₄B₂O₄
MW: 253.94 g/mol
Boiling Pt: 242.8 °C (760 mm Hg)
Melting Pt: 98…102 °C
Flash Pt: 100.7 °C
Storage Temperature: Ambient
MDL Number: MFCD06246008
CAS Number: 230299-21-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR