4,4',4'',4'''-(Pyrene-1,3,6,8-tetrayl)tetrabenzoic acid 97%

Supplier: Ambeed
Warning

A254394-1G A254394-5G A254394-100MG A254394-250MG
103812-132EA 49 USD
103812-132 103851-986 77091-426 77091-428 77238-366 77238-368
4,4',4'',4'''-(Pyrene-1,3,6,8-tetrayl)tetrabenzoic acid 97%
4,4',4'',4'''-(Pyrene-1,3,6,8-tetrayl)tetrabenzoic acid

Precaución: for Research Use Only

Formula: C₄₄H₂₆O₈
MW: 682.67 g/mol
Storage Temperature: Ambient
MDL Number: MFCD30478418
CAS Number: 933047-52-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR