meso-Tetra(4-carboxyphenyl)porphine 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
Tetrakis(4-carboxyphenyl)porphyrin, 4,4,4,4-(Porphine-5,10,15,20-tetrayl)tetrakis(benzoic acid), TCPP, Tetrakis (4-carboxyphenyl) porphyrin
A114047-100MG
A114047-250MG
A114047-25G
A114047-1G
A114047-5G
103798-338EA
18
USD
103798-338
103824-180
77025-954
77025-956
77025-958
77311-000
77311-002
meso-Tetra(4-carboxyphenyl)porphine 97%
meso-Tetra(4-carboxyphenyl)porphine
Precaución: for Research Use Only
Formula:
C₄₈H₃₀N₄O₈ MW: 790.79 g/mol Melting Pt: >300 °C Storage Temperature: Ambient |
MDL Number:
MFCD00064860 CAS Number: 14609-54-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...