4,4',4'',4'''-(Ethene-1,1,2,2-tetrayl)tetraaniline 97%
Supplier: Ambeed
|
Warning
|
A831836-1G
A831836-100MG
A831836-250MG
A831836-5G
103841-166EA
145
USD
103841-166
76845-402
77244-398
77244-400
77308-414
4,4',4'',4'''-(Ethene-1,1,2,2-tetrayl)tetraaniline 97%
4,4',4'',4'''-(Ethene-1,1,2,2-tetrayl)tetraaniline
Precaución: for Research Use Only
Formula:
C₂₆H₂₄N₄ MW: 392.49 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD30478419 CAS Number: 78525-34-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...