4,4′,4″-s-Triazine-2,4,6-triyl-tribenzoic acid 95%

Supplier: Ambeed
Warning

Synonyms: 4,4',4''-(1,3,5-Triazine-2,4,6-triyl)tribenzoic acid

A268781-25G A268781-100MG A268781-250MG
103812-932EA 28 USD
103812-932 103852-834 77092-594 77250-302 77250-304
4,4′,4″-s-Triazine-2,4,6-triyl-tribenzoic acid 95%
4,4′,4″-s-Triazine-2,4,6-triyl-tribenzoic acid

Precaución: for Research Use Only

Formula: C₂₄H₁₅N₃O₆
MW: 441.39 g/mol
Boiling Pt: 799.9 °C (760 mm Hg)
Flash Pt: 437.6 °C
Storage Temperature: Ambient
MDL Number: MFCD04116314
CAS Number: 61414-16-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR