2,2-Bis(3-amino-4-hydroxylphenyl)propane 98%

Supplier: Ambeed
Warning

A101000-100G A101000-25G A101000-5G
103795-744EA 6 USD
103795-744 103821-436 77245-096 77245-098 77282-636 77296-942
2,2-Bis(3-amino-4-hydroxylphenyl)propane 98%
2,2-Bis(3-amino-4-hydroxylphenyl)propane

Precaución: for Research Use Only

Formula: C₁₅H₁₈N₂O₂
Boiling Pt: 471 °C (760 mm Hg)
Storage Temperature: Ambient
MDL Number: MFCD00437413
CAS Number: 1220-78-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR