2,2-Bis(3-amino-4-hydroxyphenyl)hexafluoropropane 98%

Supplier: Ambeed
Warning

A308728-25G A308728-100G A308728-10G A308728-500G
103835-754EA 5 USD
103835-754 76838-514 76838-516 77250-928 77250-930 77250-932
2,2-Bis(3-amino-4-hydroxyphenyl)hexafluoropropane 98%
2,2-Bis(3-amino-4-hydroxyphenyl)hexafluoropropane

Precaución: for Research Use Only

Formula: C₁₅H₁₂F₆N₂O₂
Melting Pt: 245…248 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00039147
CAS Number: 83558-87-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR