4,4'-(1-Methylpropylidene)diphenol 97%

Supplier: Ambeed
Warning

Synonyms: Bisphenol B, 2,2-Bis(4-hydroxyphenyl)butane, 4,4'-(Butane-2,2-diyl)diphenol, 4,4'-Butane-2,2-diyldiphenol

A203830-25G A203830-5G A203830-100G A203830-10G
103848-156EA 14 USD
103848-156 77086-294 77086-296 77248-890 77248-892
4,4'-(1-Methylpropylidene)diphenol 97%
4,4'-(1-Methylpropylidene)diphenol

Precaución: for Research Use Only

Formula: C₁₆H₁₈O₂
MW: 242.32 g/mol
Boiling Pt: 412.2 °C (760 mm Hg)
Melting Pt: 126 °C
Flash Pt: 195.5 °C
Storage Temperature: Ambient
MDL Number: MFCD00020179
CAS Number: 77-40-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR