4,4'-(1,4-Phenylenebis(ethene-2,1-diyl))dibenzonitrile 97%
Supplier: Ambeed
|
Warning
|
A105392-100G
A105392-10G
A105392-25G
A105392-500G
A105392-5G
103807-928EA
292
USD
103807-928
76994-854
76994-856
76994-858
76994-860
76994-862
4,4'-(1,4-Phenylenebis(ethene-2,1-diyl))dibenzonitrile 97%
4,4'-(1,4-Phenylenebis(ethene-2,1-diyl))dibenzonitrile
Precaución: for Research Use Only
Formula:
C₂₄H₁₆N₂ MW: 332.39 g/mol Boiling Pt: 571.3 °C (760 mm Hg) Melting Pt: 230…234 °C Flash Pt: 272.8 °C Storage Temperature: Ambient |
MDL Number:
MFCD00369832 CAS Number: 13001-40-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...