4,4'-(1,2-Ethynediyl)dibenzoic Acid 98%

Supplier: Ambeed
Warning

A392045-1G A392045-100MG A392045-250MG
103826-894EA 127 USD
103826-894 76841-328 76990-142 76990-144
4,4'-(1,2-Ethynediyl)dibenzoic Acid 98%
4,4'-(1,2-Ethynediyl)dibenzoic Acid

Precaución: for Research Use Only

Formula: C₁₆H₁₀O₄
MW: 266.25 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18711382
CAS Number: 16819-43-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR