4-(Trifluoromethyl)pyrimidine-5-carboxylic acid 95%

Supplier: Ambeed
Warning

A264316-100MG A264316-250MG
103812-664EA 66 USD
103812-664 103852-556 76787-048 76787-050
4-(Trifluoromethyl)pyrimidine-5-carboxylic acid 95%
4-(Trifluoromethyl)pyrimidine-5-carboxylic acid

Precaución: for Research Use Only

Formula: C₆H₃F₃N₂O₂
Storage Temperature: Ambient
MDL Number: MFCD11106713
CAS Number: 220880-12-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR