2-Hydroxy-4-(trifluoromethyl)pyridine 95%

Supplier: Ambeed
Warning

Synonyms: 4-(Trifluoromethyl)pyridin-2-ol

A113522-100G A113522-250MG A113522-25G A113522-1G
103798-244EA 9 USD
103798-244 103824-084 77284-290 77284-292 77284-294 77310-902
2-Hydroxy-4-(trifluoromethyl)pyridine 95%
2-Hydroxy-4-(trifluoromethyl)pyridine

Precaución: for Research Use Only

Formula: C₆H₄F₃NO
MW: 163.1 g/mol
Boiling Pt: 221.2 °C (760 mm Hg)
Melting Pt: 161…165 °C
Storage Temperature: Ambient
MDL Number: MFCD00153195
CAS Number: 50650-59-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR