4-(Trifluoromethyl)-2-pyridine carboxylic acid 97%

Supplier: Ambeed
Warning

Synonyms: 4-(Trifluoromethyl)picolinic acid

A254827-1G A254827-5G A254827-100MG A254827-10G A254827-250MG
103812-170EA 19 USD
103812-170 103852-022 77091-472 77091-474 77289-120 77289-122 77289-124
4-(Trifluoromethyl)-2-pyridine carboxylic acid 97%
4-(Trifluoromethyl)-2-pyridine carboxylic acid

Precaución: for Research Use Only

Formula: C₇H₄F₃NO₂
MW: 191.11 g/mol
Melting Pt: 157…162 °C
Storage Temperature: Ambient
MDL Number: MFCD07438043
CAS Number: 588702-62-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR