4-(Trifluoromethyl)phenylacetonitrile 98%

Supplier: Ambeed
Warning

Synonyms: 4-(Trifluoromethyl)benzyl cyanide

A176043-10G A176043-25G A176043-5G
103845-426EA 20 USD
103845-426 76690-354 77082-776 77301-422
4-(Trifluoromethyl)phenylacetonitrile 98%
4-(Trifluoromethyl)phenylacetonitrile

Precaución: for Research Use Only

Formula: C₉H₆F₃N
MW: 185.15 g/mol
Boiling Pt: 246 °C (760 mm Hg)
Melting Pt: 47…49 °C
Flash Pt: 113 °C
Storage Temperature: Ambient
MDL Number: MFCD00001921
CAS Number: 2338-75-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR