4-(Trifluoromethyl)nicotinic acid 98%

Supplier: Ambeed
Warning

A114766-10G A114766-1G A114766-5G
103798-466EA 7 USD
103798-466 103824-308 77136-196 77297-322 77311-132
4-(Trifluoromethyl)nicotinic acid 98%
4-(Trifluoromethyl)nicotinic acid

Precaución: for Research Use Only

Formula: C₇H₄F₃NO₂
Boiling Pt: 290.4 °C (760 mm Hg)
Melting Pt: 146…148 °C
Flash Pt: 129.4 °C
Storage Temperature: Ambient
MDL Number: MFCD00082626
CAS Number: 158063-66-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR