4-(Trifluoromethyl)nicotinamide 98%
Supplier: Ambeed
|
Warning
|
A227974-1G
A227974-10G
A227974-250MG
A227974-5G
103819-350EA
36
USD
103819-350
103850-258
77089-046
77139-984
77139-986
77303-808
4-(Trifluoromethyl)nicotinamide 98%
4-(Trifluoromethyl)nicotinamide
Precaución: for Research Use Only
Formula:
C₇H₅F₃N₂O MW: 190.1226 g/mol Boiling Pt: 292 °C (760 mm Hg) Melting Pt: 165 °C Flash Pt: 130.4 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00126658 CAS Number: 158062-71-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...