4-(Trifluoromethyl)cyclohexanol (cis- and trans- mixture) 97%

Supplier: Ambeed
Warning

Synonyms: 4-(Trifluoromethyl)cyclohexanol, 4-(Trifluoromethyl)cyclohexan-1-ol

A135743-100MG A135743-10G A135743-250MG A135743-1G A135743-5G
103802-210EA 26 USD
103802-210 77285-490 77285-492 77285-494 77315-056 77315-058
4-(Trifluoromethyl)cyclohexanol (cis- and trans- mixture) 97%
4-(Trifluoromethyl)cyclohexanol (cis- and trans- mixture)

Precaución: for Research Use Only

Formula: C₇H₁₁F₃O
MW: 168.16 g/mol
Boiling Pt: 160.4 °C (760 mm Hg)
Storage Temperature: Ambient
MDL Number: MFCD00102144
CAS Number: 30129-18-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR