4-(Trifluoromethyl)benzoic acid hydrazide 98%

Supplier: Ambeed
Warning

Synonyms: 4-(Trifluoromethyl)benzoylhydrazine, 4-(Trifluoromethyl)benzhydrazide, 4-(Trifluoromethyl)benzoic Hydrazide, 4-(Trifluoromethyl)benzohydrazide

A204157-5G A204157-10G
103817-532EA 5 USD
103817-532 103848-194 77086-350 77237-234
4-(Trifluoromethyl)benzoic acid hydrazide 98%
4-(Trifluoromethyl)benzoic acid hydrazide

Precaución: for Research Use Only

Formula: C₈H₇F₃N₂O
MW: 204.15 g/mol
Boiling Pt: 470.6 °C (760 mm Hg)
Melting Pt: 115…119 °C at N/A mmHg
Flash Pt: 238.4 °C
Storage Temperature: Ambient
MDL Number: MFCD00051703
CAS Number: 339-59-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR