4-(Trifluoromethyl)benzamidine hydrochloride 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
4-(Trifluoromethyl)benzimidamide hydrochloride
A383732-10G
A383732-250MG
A383732-1G
A383732-25G
A383732-5G
103826-746EA
6
USD
103826-746
103837-784
76817-682
76817-684
76841-116
76841-118
76841-120
4-(Trifluoromethyl)benzamidine hydrochloride 97%
4-(Trifluoromethyl)benzamidine hydrochloride
Precaución: for Research Use Only
Formula:
C₈H₇F₃N₂·HCl MW: 224.61 g/mol Melting Pt: 158…161 °C Flash Pt: 127.7 °C Storage Temperature: Ambient |
MDL Number:
MFCD04114431 CAS Number: 38980-96-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...