4-(Trifluoromethyl)benzenesulfonyl chloride 98%

Supplier: Ambeed
Danger

A502846-10G A502846-25G A502846-5G
103839-396EA 5 USD
103839-396 76774-504 76843-366 77307-624
4-(Trifluoromethyl)benzenesulfonyl chloride 98%
4-(Trifluoromethyl)benzenesulphonyl chloride

Precaución: for Research Use Only

Formula: C₇H₄ClF₃O₂S
Boiling Pt: 252.8 °C (760 mm Hg)
Melting Pt: 30…34 °C
Flash Pt: 110 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00042422
CAS Number: 2991-42-6
UN: 3261
ADR: 8,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR