4-(Trifluoromethyl)benzenesulfonamide 97%

Supplier: Ambeed
Warning

Synonyms: α,α,α-Trifluoro-p-toluenesulphonamide

A110198-100G A110198-10G A110198-25G
103823-446EA 8 USD
103823-446 76765-758 76765-760 76765-762
4-(Trifluoromethyl)benzenesulfonamide 97%
4-(Trifluoromethyl)benzenesulphonamide

Precaución: for Research Use Only

Formula: C₇H₆F₃NO₂S
MW: 225.19 g/mol
Melting Pt: 175…180 °C
Flash Pt: 138.1 °C
Storage Temperature: Ambient
MDL Number: MFCD00159251
CAS Number: 830-43-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR