4-(Trifluoromethyl)benzamide 98%

Supplier: Ambeed
Warning

A214481-10G A214481-25G A214481-5G
103849-112EA 13 USD
103849-112 76770-646 77087-632 77303-354
4-(Trifluoromethyl)benzamide 98%
4-(Trifluoromethyl)benzamide

Precaución: for Research Use Only

Formula: C₈H₆F₃NO
MW: 189.14 g/mol
Boiling Pt: 254.2 °C (760 mm Hg)
Melting Pt: 184…186 °C
Flash Pt: 107.5 °C
Storage Temperature: Ambient
MDL Number: MFCD00007998
CAS Number: 1891-90-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR