4'-(Trifluoromethyl)acetophenone 98%

Supplier: Ambeed
Warning

A198015-10G A198015-1G A198015-500G A198015-25G A198015-5G A198015-100G
103847-610EA 14 USD
103847-610 76814-158 76814-160 76814-162 77085-626 77085-628 77550-260
4'-(Trifluoromethyl)acetophenone 98%
4'-(Trifluoromethyl)acetophenone

Precaución: for Research Use Only

Formula: C₉H₇F₃O
Boiling Pt: 79…80 °C (8 mm Hg)
Melting Pt: 30…33 °C
Flash Pt: 84.4 °C
Storage Temperature: Ambient
MDL Number: MFCD00000401
CAS Number: 709-63-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR